Pierce™ Phosphoprotein Phosphate Estimation Kit, Thermo Scientific
Supplier: 0000013509
23270
PI23270EA 734.46 USD
PI23270
Pierce™ Phosphoprotein Phosphate Estimation Kit, Thermo Scientific
Assays Protein Assays
Pierce™ Phosphoprotein Phosphate Estimation Assay Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as well as semi-quantitative assessment of the phosphorylation level.
  • Specifically measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only
  • Test tube and 96-well microplate protocols included and require less than 90 minutes to perform
  • Semi-quantitative, calculate moles of phosphate per mole of protein using the included phosvitin standard
  • Format is adaptable to development of specific assays for pure phosphoproteins when previously characterized standards are available
Applications include identification of proteins as containing phosphoserine or phosphothreonine phosphorylations; estimating the amount of pS and pT phosphorylation; developing specific quantitative assays for well-characterised phosphoproteins of interest.
Delivery information: Complete kit includes Ammonium molybdate Solution (25 mL), Malachite green solution (75 mL), Phosvitin positive control (1 mg), and TBS pack (makes 500 mL).