Pierce™ Protein A Plus Agarose, Thermo Scientific
Supplier: 0000013509
Thermo Scientific Pierce Protein A Plus agarose is a high capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
- Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein A is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
- High capacity beads with a dense load of immobilized Protein A
Caution: For Research Use Only. Not for use in diagnostic procedures.