Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Supplier: 0000013509
21050
PI21050EA 222.65 USD
PI21050
Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
Western blot signal enhancer is used to pre-treat either nitrocellulose or PVDF membranes for a 3 – 10X increase in signal intensity for most target proteins.
  • Protein targets can be detected at lower levels
  • Enhances antibody binding to immobilized antigens
  • Works with any substrate, enhancing both chemiluminescence and colorimetric detection
  • Works with any membrane
  • Ready-to-use and can be stored at room temperature
Signal enhancement has been demonstrated with targets such as IL-6, p53, NF[kappa]B, BRCA1, and EGF.