Pierce™ AminoLink™ Plus Micro Immobilization Kit, Thermo Scientific
Supplier: 0000013509
20475
PI20475EA 649.85 USD
PI20475
Pierce™ AminoLink™ Plus Micro Immobilization Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ AminoLink™ Plus Micro Immobilization Kit enables covalent attachment of amine-containing ligands for use in antibody purification and other affinity protocols.
  • Ideal for coupling small amounts (25 to 250 µg) of purified antibodies, proteins and other amine-containing ligands
  • Covalent attachment of the protein to resin results in a leach-resistant affinity support
  • Fast and easy to use – perform wash and elution steps using a microcentrifuge
  • Prepared peptide resin can be used several times with no significant loss of capacity
  • Immobilization efficiency greater than 80%
Delivery information: Kit contains AminoLink™ Plus Spin Columns (0.1 mL, 10 columns), Wash Solution (25 mL), Sodium Cyanoborohydride Solution 5M (0.5 mL), Quenching Buffer (60 mL), Elution Buffer (50 mL), PBS Pack (makes 500 mL) and Collection Tubes (200 tubes).