Pierce™ Jacalin Agarose, Thermo Scientific
Supplier: Invitrogen
20395
PI20395EA 374.4 USD
PI20395
Pierce™ Jacalin Agarose, Thermo Scientific
Electrophoresis Gels Agarose and Acrylamide
Thermo Scientific Pierce Jacalin Agarose is effective for specific affinity purification of human IgA from serum or colostrum or efficient removal of contaminating IgA from IgG samples.
  • Immobilized jacalin specifically binds human IgA1 (does not bind IgG and does not bind human IgA2 or IgA from other species)
  • Resin is loaded with approx. 4.5 mg Jacalin/mL resin and effectively binds 1 to 3 mg human IgA1/mL resin
  • Agarose resin support is 6% crosslinked beaded agarose, the most popular and versatile resin for protein affinity purification methods
Jacalin is an alpha-D-galactose binding lectin extracted from jack-fruit seeds (Artocarpus integrifolia). This lectin is a glycoprotein (approx. 40 kDa) composed of four identical subunits.
Ordering information: Upon receipt store product at 4 °C. Product is shipped at ambient temperature. Do not freeze product.
Caution: For Research Use Only. Not for use in diagnostic procedures.