Boronic Acid Resin, Pierce™
Supplier: 0000013509
Thermo Scientific Pierce Boronic Acid Resin enables ribonucleotide and oligonucleotide RNA isolation.
- Ligand: boronic acid
- Support: beaded polyacrylamide (1800 MW exclusion limit)
- Spacer: m-aminophenyl group
- Loading: 100 µMol boronate per milliliter of settled resin
- Capacity: at least 99% binding and recovery of 110 µMol AMP per milliliter of settled resin
- Format: 50% resin slurry in 0.05% sodium azide
The immobilized boronic acid is an easy-to-use polyacrylamide affinity support for purification of ribonucleotides and other small molecular weight compounds that contain cis-diol groups. Boronic acid affinity chromatography has been used to isolate ribonucleoside, to purify nucleosidyl peptide, separate ribonucleosidases in tissue extracts, separate RNA and oligoribonucleotide, and determine of the amount of non-enzymatic glycosylation present in peripheral nerve from diabetic and control rats and dogs.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.
Specification Test Results
VISUAL | Off-white gel slurry, free of foreign matter |
ASSAY: % Recovery of AMP | 95 - 105% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...