Pierce™ Recombinant C-Myc Peptide
Supplier: 0000013509
20170
PI20170EA 348.46 USD
PI20170
Pierce™ Recombinant C-Myc Peptide
Proteins and Peptides
The c-Myc peptide may be used to elute c-Myc-tagged proteins from Thermo Scientific Anti-c-Myc Agarose, when the bound protein is sensitive to conditions of chemical elution, such as extremes in pH, or sodium thiocyanate.
  • Competes out the binding of monoclonal anti-c-Myc or rabbit anti-c-Myc antibodies
  • Synthetic peptide provided as a lyophilized powder
  • Used with water to prepare stock solution
The peptide is useful for competing out anti-c-Myc antibody binding to c-Myc-tagged fusion proteins in immunoassays. The successful inhibition of antibody binding ascertains that the experimental observed binding is specific.
Caution: For Research Use Only. Not for use in diagnostic procedures.