Pierce™ Amino Acid Calibration Standard for HPLC, Thermo Scientific
Supplier: 0000013509
20088
PI20088EA 232.77 USD
PI20088
Pierce™ Amino Acid Calibration Standard for HPLC, Thermo Scientific
Standards Organic Standards
Thermo Scientific Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5 µMol/mL each in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.
  • Complete mixture—contains eighteen (18) amino acids for assessment of amino acid composition of peptide hydrolysates
  • Quantitative—contains exact amounts of each amino acid (2.5±0.1 µMol/mL, except cystine at 1.25±0.1 µMol/mL), enabling its use as a quantitation standard
  • Stable—dissolved in 0.1N hydrochloric acid and stabilized as 1 mL aliquots in sealed glass ampules
  • Standard H—designed for compositional amino acid analysis by HPLC methods of protein or peptide hydrolysates
This quantitative solution of eighteen purified amino acids in 0.1N HCl is an ideal reference standard for calibrating amino acid analyzers and assessing the composition of hydrolyzed peptides and proteins (hydrolysates). To permit standardization of microbiological and other assays, the mixture uses only the L-form of each amino acid. The molar concentration of each amino acid is verified by conventional amino acid analysis methods, making the mixture suitable as both a qualitative and quantitative standard. The solution is provided as stable 1 mL aliquots in sealed glass ampules.