Pierce™ Firefly Luciferase Glow Assay Kit, Thermo Scientific
Supplier: 0000013509
16176 16177
PI16176EA 160.8 USD
PI16176 PI16177
Pierce™ Firefly Luciferase Glow Assay Kit, Thermo Scientific
Assays Protein Assays
The Pierce™ Firefly Luciferase Glow Assay Kit provides a bright and stable bioluminescence signal (half-life greater than 2 hours) in the presence of firefly luciferase reporters for protein expression assays.
  • Highly sensitive detection of firefly luciferase activity
  • Increased signal stability
  • Contains a universal cell lysis buffer and optimised glow assay reagent
The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The Firefly Luciferase Glow Kit produces an extended light signal correlating with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.
Delivery information: Kit contains Firefly Glow Assay Buffer (5 or 50 ml), D-Luciferin (3 or 30 mg) and 2X Cell Lysis Buffer (6 or 60 ml).
Caution: For Research Use Only. Not for use in diagnostic procedures. It stored at –20 °C