Pierce QuantaBlu™ Fluorogenic Peroxidase Substrates, Thermo Scientific
Supplier: 0000013509
15169
PI15169EA
401.23
USD
PI15169
Pierce QuantaBlu™ Fluorogenic Peroxidase Substrates, Thermo Scientific
Electrophoresis Reagents
Western Blotting Reagents
The substrate generates a blue fluorescent product upon reaction with peroxidase and does not photobleach.
- More sensitive than TMB, OPD or ABTS substrates
- Large dynamic range (4 log peroxidase concentration range)
- Working solution stable for 24 hours
- Large stokes shift; excitation/emission maxima of 325/420; range of 315 to 345/370 to 460
- Does not photobleach
QuantaBlu™ substrate also allows for stopped, nonstopped, and kinetic assays to be performed with low-level detection sensitivity and high signal-to-noise ratios.
Delivery information: Kit includes QuantaBlu™ substrate solution (250 mL), QuantaBlu™ peroxide solution (30 mL), QuantaBlu stop solution (275 mL).
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)