Pierce™ North2South™ Hybridization Buffer, Thermo Scientific
Supplier: 0000013509
37549
PI-37549EA 315.38 USD
PI-37549
Pierce™ North2South™ Hybridization Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Hybridization Buffers
Thermo Scientific North2South Hybridization Buffer is an aqueous buffer, pH 6.9, for use with the North2South Chemiluminescent Detection Kit.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8˚C.