Esteem® CP Sterile Synthetic Gloves, Cardinal
Supplier: Cardinal Health
2Y72PL55 2Y72PL60 2Y72PL65 2Y72PL70 2Y72PL75 2Y72PL80 2Y72PL85 2Y72PL90
PH2Y72PL55CS 1526.85 USD
PH2Y72PL55 PH2Y72PL60 PH2Y72PL65 PH2Y72PL70 PH2Y72PL75 PH2Y72PL80 PH2Y72PL85 PH2Y72PL90
Esteem® CP Sterile Synthetic Gloves, Cardinal
Gloves
Hand-specific, powder-free clean process gloves are made of polyisoprene synthetic polymer for a fit, feel, and performance that is virtually indistinguishable from latex.
  • Superior protection against bacteria and particulates
  • Highly breathable and fluid-resistant
  • Low particle shedding
  • Soft and hypoallergenic
  • Malleable noseband ensures custom, secure seal
These gloves contain no natural rubber latex proteins or allergens. Beaded cuff. Bisque finish. Sterilized by gamma radiation to a Sterility Assurance Level (SAL) of 10-6. Color: white.
Certifications: CE marked.
Ordering information: Individually pair packed, 50 pair/double poly bag, 200 pair/case.