Protegrity® CP Sterile Latex Gloves with Nitrile, Cardinal
Supplier: Cardinal Health
2Y72N1 2Y72N2 2Y72N3 2Y72N4 2Y72N5 2Y72N6 2Y72N7
PH2Y72N0CS 1073.3 USD
PH2Y72N0 PH2Y72N1 PH2Y72N2 PH2Y72N3 PH2Y72N4 PH2Y72N5 PH2Y72N6 PH2Y72N7
Protegrity® CP Sterile Latex Gloves with Nitrile, Cardinal
Gloves
Triple-layer, powder-free, clean process gloves.
  • Triple layer, powder-free
  • Comfort of natural rubber latex with the strength of nitrile for exceptional grip and tactile properties
  • Hand specific, beaded cuff, bisque finish
  • Sterilized by gamma irradiation to a Sterility Assurance Level 10⁻⁶
Unique bonding technology integrates the comfort of natural rubber latex with the strength of nitrile for an exceptional grip, outstanding tactile properties, and enhanced comfort. The natural rubber latex outer layer provides elasticity and tensile strength.
The synthetic nitrile inner layer, closest to the skin, enables effortless donning as well as durability. The middle layer is a blend of natural rubber and synthetic nitrile formulated to bond the inner and outer layers. Hand-specific. Beaded cuff. Bisque finish.
Certifications: CE marked.
Ordering information: 50 individually packed pairs per double poly bag, 200 pairs per case.
Caution: Safe use of this glove by or on latex-sensitized individuals has not been established.