CP100™ Nitrile Gloves, Cardinal
Supplier: Cardinal Health
2Y1851 2Y1852 2Y1849 2Y1850 2Y1853
PH2Y1851CS 1171.71 USD
PH2Y1851 PH2Y1852 PH2Y1849 PH2Y1850 PH2Y1853
CP100™ Nitrile Gloves, Cardinal
Gloves
Clean-processed nitrile gloves offer excellent strength and a comfortable fit.
  • Clean processed, powder free
  • Excellent strength, textured grip, comfortable fit
  • Ambidextrous, beaded cuff
ESD properties and 29.2 cm (11¹/₂") length make gloves ideal for many cleanroom applications. Textured surface provides a good grip. Beaded cuff to prevent rolldown. Color: white.
Ordering information: Packaged 100 gloves per double poly bag, 1000 gloves per case.