CP100™ BT Latex Ambi Gloves, Cardinal

Supplier: Cardinal Health
2Y1731 2Y1732 2Y1733 2Y1730
PH2Y1731CS 1125.5 USD
PH2Y1731 PH2Y1732 PH2Y1733 PH2Y1730
CP100™ BT Latex Ambi Gloves, Cardinal
Gloves
Ambidextrous gloves feature a beaded cuff to minimize rolldown and make donning easy.

  • Clean-processed, powder-free
  • Color: natural white

Fully textured palm and fingers provide a secure grip.

Packaging: Packaged in double poly bags.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR