Multi-Flex® Clean Class 100™ Latex Gloves, Cardinal

Supplier: Cardinal Health
2Y1600 2Y1601 2Y1602 2Y1603 2Y1604 2Y1605 2Y1606 2Y1607
PH2Y1600CS 821.14 USD
PH2Y1600 PH2Y1601 PH2Y1602 PH2Y1603 PH2Y1604 PH2Y1605 PH2Y1606 PH2Y1607
Multi-Flex® Clean Class 100™ Latex Gloves, Cardinal
Gloves
Powder-free latex gloves are clean-processed to protect sensitive environments from contamination by particulate matter

  • Powder free, clean processed to protect sensitive environments
  • Hand specific, tapered cuffs
  • Anti-slip finish provides a sure grip
  • Tapered cuffs
  • Color: natural

Hand-specific gloves are tough, flexible, dependable, and strong. Thin fingertips ensure optimum fit, sensitivity, and dexterity required for small parts manufacturing procedures.

Suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.

Ordering information: Multilevel bag-in-a-bag packaging, 50 pair per bag. Case totals reflect glove pairs (case of 400 includes 200 glove pairs).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR