PolyATtract mRNA Isolation System III with Magnetic Stand, 15 isolations, Promega
Supplier: Promega Corporation
Z5300
PAZ5300EA 348.12 USD
PAZ5300
PolyATtract mRNA Isolation System III with Magnetic Stand, 15 isolations, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The PolyATtract System isolates messenger RNA directly from total RNA.
  • Isolates Highly Pure mRNA from Total RNA
  • Protocol complete in approximately 45 minutes
  • Isolated mRNA is suitable for use in in vitro translation, RT-PCR and cDNA synthesis
  • Cat.# Z5200 and Z5210 contain sufficient reagents for 3 separate mRNA isolations, each from 1-5mg total RNA
  • Cat.# Z5300 and Z5310 contain sufficient reagents for 15 separate mRNA isolations, each from 100-1,000µg total RNA
The PolyATtract mRNA Isolation Systems use the MagneSphere technology to isolate mRNA rapidly and effectively from total RNA. The systems use a biotinylated oligo(dT) primer to hybridize, at high efficiency in solution, to the 3' poly(A)+ region present in most mature eukaryotic mRNAs. The hybrids are bound to streptavidin coupled to paramagnetic particles, captured using a magnetic separation stand and washed at high stringency. The mRNA is eluted from the solid phase by the simple addition of ribonuclease-free, deionized water. With total RNA as the starting material, poly(A)+ mRNA is isolated in approximately 45 minutes. The isolated mRNA is suitable for all molecular biology applications, including in vitro translation and cDNA synthesis. Cat.# Z5200 contains sufficient reagents for 3 separate mRNA isolations, each from 1-5mg of total RNA. Cat.# Z5210 contains the same reagents as Cat.# Z5200, excluding the Magnetic Separation Stand. Cat.# Z5300 contains sufficient reagents for 15 separate mRNA isolations, each from 100-1,000ug of total RNA. Cat.# Z5310 contains the same reagents as Cat.# Z5300, excluding the Magnetic Separation Stand.