Tween® 20 (Polysorbate)

Supplier: Promega Corporation

Synonyms: Polyoxyethylene sorbitan monolaurate

W3831
PAW3831EA 58.18 USD
PAW3831
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)

Tween® 20 is a nonionic detergent used as a buffer component for immunoscreening in the ProtoBlot Systems.


Tween® 20 (100%) is used in immunoscreening to dissociate nonspecific interactions with an antibody probe.

Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR