MagnaBot Spacer ¹/₈", 1 each, Promega
Supplier: Promega Corporation
V8581
PAV8581EA 54.2 USD
PAV8581
MagnaBot Spacer ¹/₈", 1 each, Promega
Racks Magnetic Separation Racks
The spacers adjust the height of a 96-well plate placed onto the MagnaBot 96 Magnetic Separation Devices to optimize magnetization.
  • Adjust height of 96-well plate placed onto MagnaBot(R) 96 Magnetic Separation Devices
  • Optimize magnetization while using MagnaBot(R) 96 Magnetic Separation Devices
The MagnaBot Spacers are designed to optimize use of the MagnaBot 96 Magnetic Separation Device (Cat.# V8151) and the Deep Well MagnaBot 96 Magnetic Separation Device (Cat.# V3031). The spacers adjust the height of a 96-well plate placed onto the MagnaBot 96 Magnetic Separation Devices to optimize magnetization. Refer to specific robotic protocols and applications for recommended uses.