MagnaBot II Magnetic Separation Device, Promega
Supplier: Promega Corporation
V8351
PAV8351EA 1104.99 USD
PAV8351
MagnaBot II Magnetic Separation Device, Promega
Racks Magnetic Separation Racks
The MagnaBot II Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles with a 96-well PCR plate.
  • Designed to work with a 96-well PCR plate
  • Allows the quantitative removal of liquids
The MagnaBot II Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles with a 96-well PCR plate. A 96-well PCR plate containing MagneSil Paramagnetic Particles is placed on the unit to draw the particles to the side and away from the bottom of each well. This allows the quantitative removal of liquids.