MagnaBot 96 Magnetic Separation Device, Promega
Supplier: Promega Corporation
V8151
PAV8151EA 1055.02 USD
PAV8151
MagnaBot 96 Magnetic Separation Device, Promega
Racks Magnetic Separation Racks
The MagnaBot 96 Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles.
  • Designed for high-throughput bioseparation using magnetic particles
  • Compatible with many plates
The MagnaBot 96 Magnetic Separation Device is designed for high-throughput bioseparation using magnetic particles such as MagneSil Paramagnetic Particles, which use the principle of magnetic separation as an alternative to vacuum filtration and centrifugation separation formats. This device is compatible with Collection Plates (Cat.# A9161), 1.2ml, Round-Bottom Deep Well Plates (Cat.# V6771) and Greiner 96-well P.S. V-bottom plates (Greiner Cat.# 651101). A generic 96-well plate may not be compatible.