Sodium dodecyl sulfate (SDS) solution 10% (w/v), Molecular Biology Grade

Supplier: Promega Corporation

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

V6553 V6551
PAV6553EA 247.24 USD
PAV6553 PAV6551
Sodium dodecyl sulfate (SDS) solution 10% (w/v), Molecular Biology Grade
Sodium dodecyl sulphate (SDS)

SDS is a detergent that is known to denature proteins. It is used in polyacrylamide gel electrophoresis for the determination of protein molecular weight.


  • Quality tested and certified free of DNase and RNase activity


It is also used in nucleic acid extraction procedures for the disruption of cell walls and dissociation of nucleic acid:protein complexes.


A260: ≤0.3
A280: ≤0.2

Formula: CH₃(CH₂)₁₁OSO₃Na
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR