TE Buffer, 1X, Molecular Biology Grade, Promega
Supplier: Promega Corporation
TE Buffer, 1X, Molecular Grade (pH 8.0), is a buffer composed of 10 mM Tris-HCl containing 1 mM EDTA.Na2.
TE Buffer is free of DNase, Rnase, and protease activity.
At 25 °C, its pH is 7.9 - 8.1.
A280: ≤0.05