TBE Buffer, 10X, Molecular Biology Grade, Promega
Supplier: Promega Corporation
V4251
PAV4251EA 88.31 USD
PAV4251
TBE Buffer, 10X, Molecular Biology Grade, Promega
Buffers Electrophoresis Buffers Running Buffers
TBE Buffer, 10X (pH 8.3), is used for polyacrylamide and agarose gel electrophoresis.
  • Quality tested; demonstrates ≤1% release of DNase and RNase activity
This product has been optimized for use in DNA applications.