X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 50 mg/mL in DMF

Supplier: Promega Corporation
Danger

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

V3941
PAV3941EA 115.16 USD
PAV3941
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 50 mg/mL in DMF
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

X-Gal, in conjunction with IPTG, is used to detect β-galactosidase activity to differentiate recombinants from non-recombinants in cloning experiments using vectors containing the lacZ or lacZ α-peptide gene.


  • Quality tested for use with the pGEM®-Z Vectors in a chromogenicity assay
  • Supplied at a concentration of 50mg/ml in dimethylformamide, 2.0ml/vial


This item has a concentration of 50 mg/mL in dimethylformamide, 2.0 mL/vial.
Storage: -30°C to -10°C

Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8
UN: 2265
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR