Bisacrylamide ≥99.0%, Molecular Grade

Supplier: Promega Corporation
Warning

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

V3143
PAV3141EA 41.23 USD
PAV3141 PAV3143
Bisacrylamide ≥99.0%, Molecular Grade
Bisacrylamide

Bisacrylamide, Molecular Grade, is a cross-linking agent used in the preparation of polyacrylamide gels. This product is tested for its efficiency in gel polymerization.


  • Quality tested and certified free of DNase, RNase and protease activity


Form: White, free-flowing crystals.
Storage: 15°C to 30°C

Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Boiling Pt: 125
Melting Pt: 185
Density: 1.235 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99.0%
Acrylic Acid (CH2:CHCOOH) ≤0.001%
A290 (1% solution) ≤0.20
Magnesium ≤2 ppm
Conductivity (2% in water) ≤10 μmhos


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR