dTTP, 40 µmol, Promega
Supplier: Promega Corporation
U1231
PAU1231EA 131.75 USD
PAU1231
dTTP, 40 µmol, Promega
Nucleic Acid Reagents Nucleotides
High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.
  • High-Quality Nucleotides for Your Workflow
  • Greater than 99% triphosphate content
  • 100mM in water at pH 7.5
High-quality deoxynucleotide triphosphates (dNTPs) are critical for the success of many key procedures such as cDNA synthesis, sequencing and labeling. Promega dNTPs have greater than 99% triphosphate content and are provided at a concentration of 100mM in water at pH 7.5.