Western Blue Stabilized Substrate for Alkaline Phosphatase, 100 ml, Promega
Supplier: Promega Corporation
S3841
PAS3841EA 82.13 USD
PAS3841
Western Blue Stabilized Substrate for Alkaline Phosphatase, 100 ml, Promega
Electrophoresis Reagents Western Blotting Reagents
Stable, ready-to-use substrate for Western blots and immunoscreening.
  • Ready-to-use formulation that does not require dilution or reagent mixing
  • As sensitive as other commercially available BCIP/NBT formulations and reagents
  • Stable for one year at room temperature
Western Blue Stabilized Substrate for Alkaline Phosphatase is a stable, ready-to-use substrate for Western blots and immunoscreening. It is a mixture of 5-bromo-4-chloro-3-indolyl-phosphate (BCIP) and nitro blue tetrazolium (NBT) in a proprietary stabilizing buffer. Western Blue Substrate should be used directly and without dilution. This liquid substrate deposits a permanent dark purple stain on membrane sites bearing alkaline phosphatase. Western Blue Substrate is as sensitive as other reagents based on the BCIP/NBT formulation.
This liquid substrate deposits a permanent dark purple stain on membrane sites bearing alkaline phosphatase. Western Blue Substrate is as sensitive as other reagents based on the BCIP/NBT formulation.
Western Blue Substrate should be used directly and without dilution.