AttoPhos Buffer, Promega
Supplier: Promega Corporation
S1021
PAS1021EA
74.52
USD
PAS1021
AttoPhos Buffer, Promega
Buffers
Assay Buffers
The AttoPhos(R) Buffer is optimized for sensitive detection of alkaline phosphatase (AP) when used to prepare working solutions with AttoPhos(R) Substrate (Cat.# S1011, S1012, S1013).