MULTI-CORE™ Buffer Pack, Promega
Supplier: Promega Corporation
Universal restriction enzyme buffer. Supports double-digests with many restriction enzymes.
- Convenient Aliquots of the Promega Universal Restriction Enzyme 10X Buffer
- Enables digestion of DNA with more than one enzyme in a single reaction
- Only modest adjustments in the amount of enzyme used will ensure complete multiple digestions
The MULTI-CORE™ Buffer is formulated to provide simple buffering conditions for performing multiple digestions.
MULTI-CORE™ Buffer (1X) = 25mM Tris-acetate (pH 7.5 at 37°C), 100mM potassium acetate, 10mM magnesium acetate, 1mM DTT.
Many Promega restriction enzymes have between 50% and 100% activity in reactions using this buffer.
Caution: For laboratory use. Outside of the United States, this product is intended for research use only unless otherwise stated.