Nhe I (Genome Qualified)
Supplier: Promega Corporation
R6501
R6505
PAR6501EA
124.52
USD
PAR6501
PAR6505
Nhe I (Genome Qualified)
Nucleic Acid Reagents
Restriction Enzymes
G/CTAG C C GATC\G
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Genome qualified to ensure optimal performance in genomic analysis applications