Nhe I (Genome Qualified)
Supplier: Promega Corporation
R6501 R6505
PAR6501EA 124.52 USD
PAR6501 PAR6505
Nhe I (Genome Qualified)
Nucleic Acid Reagents Restriction Enzymes
G/CTAG C C GATC\G
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Genome qualified to ensure optimal performance in genomic analysis applications