SP6 Promoter Primer, 2 µg, Promega
Supplier: Promega Corporation
Q5011
PAQ5011EA
120.76
USD
PAQ5011
SP6 Promoter Primer, 2 µg, Promega
Nucleic Acid Reagents
Primers and Probes
The SP6 Promoter Sequencing Primer is used for sequencing inserts cloned into various vectors.
- Designed for Sequencing Inserts Cloned into the pGEM(R), pALTER(R)-MAX and pCI-neo Vectors
- Designed to be annealed to single-stranded DNA or, after alkaline denaturation, to double-stranded DNA
- Purified by gel electrophoresis or HPLC
- Primer Sequence: 5´-d(TATTTAGGTGACACTATAG)-3´
- Supplied at a concentration of 10µg/ml
The SP6 Promoter Primer is designed for sequencing inserts cloned into the pGEM, pALTER-MAX and pCI-neo Vectors. The primer is designed to be annealed to single-stranded DNA or, after alkaline denaturation, to double-stranded DNA. The promoter primer is purified by gel electrophoresis or HPLC. Primer Sequence: 5'-d(TATTTAGGTGACACTATAG)-3'.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...