SP6 RNA Polymerase (HC), 2,500 u, Promega
Supplier: Promega Corporation
P4084
PAP4084EA 949.62 USD
PAP4084
SP6 RNA Polymerase (HC), 2,500 u, Promega
Nucleic Acid Reagents RNA Synthesis and Transcription
DNA-dependent RNA polymerase for in vitro transcription.
  • DNA-Dependent RNA Polymerase Specific for SP6 Promoter Sequences
  • Exhibits extremely high affinity and specificity for SP6 promoter sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (
SP6 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only SP6 DNA or DNA cloned downstream from an SP6 promoter can serve as a template for SP6 RNA Polymerase-directed RNA synthesis.