T7 RNA Polymerase (HC), Promega
Supplier: Promega Corporation
P4074
PAP4074EA 973.72 USD
PAP4074
T7 RNA Polymerase (HC), Promega
Nucleic Acid Reagents RNA Synthesis and Transcription
DNA-dependent RNA polymerase for in vitro transcription.
  • DNA-Dependent RNA Polymerase Specific for T7 Promoter Sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (
  • Incorporates 32P, 33P, 3H and 35S nucleoside triphosphates
T7 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only T7 DNA or DNA cloned downstream from a T7 promoter can serve as a template for T7 RNA Polymerase-directed RNA synthesis.