T3 RNA Polymerase (HC), 2500 U, Promega
Supplier: Promega Corporation
P4024
PAP4024EA 219.8 USD
PAP4024
T3 RNA Polymerase (HC), 2500 U, Promega
Nucleic Acid Reagents RNA Synthesis and Transcription
DNA-dependent RNA polymerase for in vitro transcription.
  • DNA-Dependent RNA Polymerase Specific for T3 Promoter Sequences
  • Exhibits extremely high affinity and specificity for T3 promoter sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (
  • Supplied at a concentration of 10-20u/microl (Cat.# P2083) and 80u/µl (Cat.# P4024)
T3 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only T3 DNA or DNA cloned downstream from a T3 promoter can serve as a template for T3 RNA Polymerase-directed RNA synthesis.