pGEM Express Positive Control Template, 10 µg, Promega
Supplier: Promega Corporation
P2561
PAP2561EA 83.44 USD
PAP2561
pGEM Express Positive Control Template, 10 µg, Promega
Nucleic Acid Reagents RNAi
The pGEM Express Positive Control Template is used to monitor in vitro transcription reactions when using the Riboprobe Systems.
  • Used with Riboprobe(R) Systems to Monitor in vitro Transcription Reactions
  • Can be used with SP6, T7 or T3 RNA polymerase
  • SP6 RNA polymerase produces transcripts of 1,787 and 2,566 bases; T7 RNA polymerase produces transcripts of 1,065 and 2,346 bases; T3 RNA Polymerase produces transcripts of 250 and 1,525 bases
The pGEM Express Positive Control Template is created by linearizing a vector with the restriction enzyme ScaI. The Positive Control Template may be used to monitor in vitro transcription reactions when using the Riboprobe Systems.
Caution: Store at –20°C