Dithiothreitol (DTT, Cleland's reagent) 100 mM in water ≥99.0%, Molecular Grade

Supplier: Promega Corporation

P1171
PAP1171EA 25.09 USD
PAP1171
Dithiothreitol (DTT, Cleland's reagent) 100 mM in water ≥99.0%, Molecular Grade
Dithiothreitol (DTT, Cleland's reagent)

DTT, Molecular Grade, is an antioxidant used to stabilize enzymes and other proteins containing sulfhydryl groups. The liquid form of the product is a 100mM solution of DTT in water.


  • Quality tested and certified free of DNase, RNase and protease activity
  • Available as a liquid (100µl) or as a powder (5g or 25g)
  • Supplied at a concentration of 100mM


Physical/Chemical Properties of Powder:
Form: White crystals/powder
Purity: ≥99.0%
Melting Point: 40 to 44°C
A283 at 20 mM: ≤0.04
Oxidized at ≤0.50%

Storage: -30°C to -10°C

Formula: C₄H₁₀O₂S₂
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR