MagneSil ONE, Fixed Yield Blood Genomic System, 1 x 96 preps, Promega
Supplier: Promega Corporation
MD1370
PAMD1370PK 503.56 USD
PAMD1370
MagneSil ONE, Fixed Yield Blood Genomic System, 1 x 96 preps, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The MagneSil ONE, Fixed Yield Blood Genomic System purifies 1ug of DNA (+/- 50%) from 60ul of anti-coagulated whole blood.
  • Automated Extraction of 1µg DNA from 60µl Blood
  • Process 96 samples in 1 hour with no hands-on time following robot setup
  • DNA suitable for use in PCR, multiplex PCR and SNP genotyping applications
  • Validated automated methods available
The MagneSil ONE, Fixed Yield Blood Genomic System purifies 1ug of DNA (+/- 50%) from 60ul of anti-coagulated whole blood. Purification of a fixed yield of DNA eliminates the need to quantitate and normalize concentrations postpurification. The highly pure DNA isolated is suitable for use in PCR, multiplex PCR and SNP genotyping applications. Walkaway automation is available on the Beckman Coulter Biomek FX in a 96-well format. Process 96 samples in about 1 hour with no hands-on time following robot setup.