MagneSil Blood Genomic, Max Yield System, 1 x 96 preps, Promega
Supplier: Promega Corporation
MD1360
PAMD1360PK 429.4 USD
PAMD1360
MagneSil Blood Genomic, Max Yield System, 1 x 96 preps, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MagneSil Blood Genomic, Max Yield System provides automated high-throughput DNA purification on the Beckman Coulter Biomek FX using MagneSil Paramagnetic Particle technology.
  • Walkaway Automation of Genomic DNA Extraction
  • Purifies DNA from 96 samples of anti-coagulated whole blood in 1 1/2 hours
  • Average yield of 96 samples from normal healthy adults is ≥4microg
  • Validated automated methods available
The MagneSil Blood Genomic, Max Yield System provides automated high-throughput DNA purification on the Beckman Coulter Biomek FX using MagneSil Paramagnetic Particle technology. DNA from 96 samples of anti-coagulated human whole blood is purified in about 1 1/2 hours with no hands-on time once the robot protocol is initiated. Studies on DNA recovery and purity and PCR results show no cross-contamination between samples in adjacent wells. Purified DNA is qualified for single-locus simple PCR as well as more demanding applications such as multiplex PCR (e.g., PowerPlex 16 System [Cat.# DC6531], Y Chromosome Deletion Detection System [Cat.# MD1531]) and SNP genotyping.