M-MLV Reverse Transcriptase, RNase H Minus, Promega
Supplier: Promega Corporation
M5301
PAM5301EA 292.48 USD
PAM5301
M-MLV Reverse Transcriptase, RNase H Minus, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Deletion mutation of M-MLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.
  • Helpful for Preparing Long cDNAs and Libraries with a High Percentage of Full-Length cDNA
  • Supplied at a concentration of 100-200u/microl
Moloney Murine Leukemia Virus Reverse Transcriptase, RNase H Minus (M-MLV RT [H-]), is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb). This form of M-MLV Reverse Transcriptase has been genetically altered to remove the associated RNase H activity. Although many researchers are successful in using M-MLV RT (H+) for analytical and some preparative cDNA applications, reverse transcriptases lacking RNase H activity provide another option to prepare long cDNAs and libraries containing a high percentage of full-length cDNA.
Caution: Storage temperature -30°C to -10°C