CIAP Buffer Pack, Promega
Supplier: Promega Corporation
M1833
PAM1833EA 34.12 USD
PAM1833
CIAP Buffer Pack, Promega
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
A complete system for phosphorylating both double- and single-stranded DNA and RNA with T4 PNK and [gamma-32P]ATP.
  • Phosphorylates RNA, ssDNA and dsDNA Ends with a Purchased Radiolabeled Nucleotide
  • System includes enzymes, buffers and control DNA standards for measuring reaction efficiencies (except radionucleotides)
  • Flexible labeling works with [γ-32P]ATP, [γ-33P]ATP or [γ-35S]ATP
The system includes enzymes, buffers and control DNA standards to measure reaction efficiencies. Calf Intestinal Alkaline Phosphatase is included for removal of the 5'-phosphate prior to labeling with T4 Polynucleotide Kinase.
Caution: Storage Temperature -30°C to -10°C