Luciferase SPG/T7 Control DNA, Promega
Supplier: Promega Corporation
L4821
PAL4821EA 162.55 USD
PAL4821
Luciferase SPG/T7 Control DNA, Promega
Protein Expression Protein Expression Systems
Luciferase SP6 and T7 Control DNAs are used with the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter.
  • Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay
The Luciferase SP6 and T7 Control DNAs are used as functional controls in the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter. All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are monitored easily by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components. Luciferase Control DNAs are supplied as 0.5mg/ml solutions in TE buffer.
Luciferase Control DNAs are supplied as 0.5 mg/mL solutions in TE buffer.
All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are easily monitored by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components.