Luciferase SP6 Control DNA, Promega
Supplier: Promega Corporation
L4741
PAL4741EA 153.64 USD
PAL4741
Luciferase SP6 Control DNA, Promega
Protein Expression Protein Expression Systems
Luciferase SP6 and T7 Control DNAs are used with the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter.
  • Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay
The Luciferase SP6 and T7 Control DNAs are used as functional controls in the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter. All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are monitored easily by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components. Luciferase Control DNAs are supplied as 0.5mg/ml solutions in TE buffer.