Ethidium bromide 10 mg/mL in aqueous solution for molecular biology

Supplier: Promega Corporation
Warning

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

H5041
PAH5041EA 58.71 USD
PAH5041
Ethidium bromide 10 mg/mL in aqueous solution for molecular biology
Ethidium bromide

Ethidium Bromide Solution, Molecular Grade (10mg/ml), is a fluorescent dye suitable for staining nucleic acids after electrophoresis or in cesium chloride gradients. The solution can be used to detect both double-stranded and single-stranded DNA.

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR