EDTA disodium salt dihydrate ≥99.0% for molecular biology

Supplier: Promega Corporation

H5032
PAH5032EA 140.87 USD
PAH5032
EDTA disodium salt dihydrate ≥99.0% for molecular biology
EDTA disodium salt dihydrate

EDTA, Disodium Salt, Molecular Biology Grade, is a chelator of divalent metal cations.


  • Quality tested and certified free of DNase, RNase and protease activity

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99.0%
Insolubles ≤0.005%
Lead ≤5ppm
Iron ≤10ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR