Diamond™ Nucleic Acid Dye
Supplier: Promega Corporation
H1181
PAH1181EA 106.12 USD
PAH1181
Diamond™ Nucleic Acid Dye
Stains Electrophoresis Stains
Diamond Nucleic Acid Dye is a sensitive fluorescent dye that binds to single-stranded DNA, double-stranded DNA and RNA, and can be used to stain and visualize nucleic acids in gels.
  • Detect and visualise small amounts of nucleic acids after gel electrophoresis
  • Store at room temperature for quick and easy use without thawing
  • Use with a variety of common gel types and imaging equipment
  • Concentrated dye stable for up to 90 days at room temperature
Diamond Nucleic Acid Dye is a sensitive fluorescent dye that binds to single-stranded DNA, double-stranded DNA and RNA, and can be used to stain and visualize nucleic acids in gels. Diamond Nucleic Acid Dye is compatible with denaturing and native agarose and polyacrylamide gels and can be imaged with any standard imaging system, such as by UV transillumination with a Polaroid or digital camera, GE ImageQuant or Bio-Rad Gel Doc systems. The concentrated dye is stable for up to 90 days at room temperature. Diamond Nucleic Acid Dye does not require prewashing or destaining of gels. It is more much more sensitive than ethidium bromide, so less sample nucleic acid and nucleic acid markers are required for visualization, resulting in increased savings with every gel you run.