GloMax Discover System, Promega
Supplier: Promega Corporation
GM3000
PAGM3000EA 43183.49 USD
PAGM3000
GloMax Discover System, Promega
Microplate Readers
GloMax Discover is a high-performance microplate reader developed to provide advanced detection capabilities with Promega luminescence and fluorescence based reagents.
  • Easy-to-Use Microplate Reader with Advanced Detection Capabilities
  • Luminescence, fluorescence, UV-visible absorbance, BRET and FRET detection
  • 6- to 384-well microplate reader
  • Optimized with Promega cell viability and genetic reporter assays
The GloMax Discover System is a high-performance multimode detection instrument developed with Promega reagent chemistries to provide a simple means of detecting advanced chemistries. This instrument provides researchers superior luminescence sensitivity and dynamic range for both strong and weak bioluminescence experimental samples, as well as a seamless integration with Promega’s industry leading bioluminescent assays. GloMax Discover also provides flexible use of filters for fluorescence intensity, BRET, FRET, filtered luminescence and UV-visible absorbance measurements. The instrument is operated by an integrated Tablet PC, which provides quick and easy navigation through the control options. Exporting your results is made seamless with a variety of options, including exporting to your local data network. The GloMax Discover software provides the required technical elements of a 21 CFR Part 11-compliant system to be used with the appropriate laboratory workflow.