CellTiter-Blue Cell Viability Assay, Promega
Supplier: Promega Corporation
G8082 G8080 G8081
PAG8082EA 4505.84 USD
PAG8082 PAG8080 PAG8081
CellTiter-Blue Cell Viability Assay, Promega
Assays Cellular Assays
Homogeneous, fluorometric cell viability assay (resazurin). Requires a 1- to 4-hour incubation.
  • A Homogeneous, Fluorescent Assay to Monitor Cell Viability
  • Fast, flexible, time-saving
  • Amenable to multiplexing with other assays
The CellTiter-Blue Cell Viability Assay provides a homogeneous, fluorescent method for monitoring cell viability. The assay is based on the ability of living cells to convert a redox dye (resazurin) into a fluorescent end product (resorufin). Nonviable cells rapidly lose metabolic capacity and thus do not generate a fluorescent signal. The homogeneous assay procedure involves adding the single reagent directly to cells cultured in serum-supplemented medium. After an incubation step, data are recorded using either a plate-reading fluorometer (preferred) or spectrophotometer.