BenchTop pGEM DNA Markers, 250 µl, Promega
Supplier: Promega Corporation
G7521
PAG7521EA 310.65 USD
PAG7521
BenchTop pGEM DNA Markers, 250 µl, Promega
Electrophoresis Reagents Molecular Weight Markers
36-2,645bp; 15 bands. Can be stored at room temperature. Contains dye marker, ready to load. Combination of three separate digests of pGEM(R)-3 Vector.
  • Fifteen DNA Fragments of 36-2,645bp, Combined From Three Separate Digests of pGEM(R)-3 Vector
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 5microl/lane
The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. Fifteen phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 36bp to 2,645bp. These unique markers are generated from separate digests of pGEM(R)-3 Vector DNA with HinfI, RsaI and SinI later combined to form the markers. Load 5microl/lane.